Quinoline-N-oxide hydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Quinoline 1-oxide hydrate

15343-0050 15343-0250
TS15343-0050EA 66 USD
TS15343-0050 TS15343-0250
Quinoline-N-oxide hydrate 98%
Quinoline-N-oxide hydrate
Formula: C₉H₇NO·xH₂O
MW: 145.16 g/mol (anhy)
Melting Pt: 48…53 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00149472
CAS Number: 64201-64-5

Order Now

Specification Test Results

Appearance Ochre to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 48 to 53 °C
Water ≤22.5 % (K.F.)
Titration with HClO4 ≥97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR