Rhodanine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one

15260-0250 15260-1000
TS15260-0250EA 53 USD
TS15260-0250 TS15260-1000
Rhodanine 99%
Rhodanine
Formula: C₃H₃NOS₂
MW: 133.19 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005488
CAS Number: 141-84-4
EINECS: 205-505-1

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 167 to 170 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR