1-Benzylimidazole 99%

Supplier: Thermo Scientific Chemicals

15225-0050 15225-0250 15225-1000
TS15225-0050EA 37 USD
TS15225-0050 TS15225-0250 TS15225-1000
1-Benzylimidazole 99%
1-Benzylimidazole
Formula: C₁₀H₁₀N₂
Boiling Pt: 310 °C (760 mmHg)
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00005296
CAS Number: 4238-71-5
EINECS: 224-200-4

Order Now

Specification Test Results

Appearance Brown Crystals
Infrared spectrum Conforms
Melting point 68 to 71 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR