2-Chloro-6-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-2-picoline

15223-0010 15223-0100
TS15223-0010EA 14 USD
TS15223-0010 TS15223-0100 TS15223-0500
2-Chloro-6-methylpyridine 98%
2-Chloro-6-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64
Density: 1.16 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5290 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR