2-Amino-3-nitropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine

15112-0050 15112-0250
TS15112-0050EA 41 USD
TS15112-0050 TS15112-0250
2-Amino-3-nitropyridine 99%
2-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 165.9…166.4 °C
Flash Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006314
CAS Number: 4214-75-9
EINECS: 224-144-0

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 164 to 168 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR