4-Ethylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

15057-0250 15057-1000 15057-5000
TS15057-0250EA 36 USD
TS15057-0250 TS15057-1000 TS15057-5000
4-Ethylpyridine 98%
4-Ethylpyridine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 168.3 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.942 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00006450
CAS Number: 536-75-4
EINECS: 208-646-7
UN: 1992
ADR: 3,III
Merck Index: 13,03880

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5009 to 1.5029 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR