2-Methylbenzothiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

15038-0250 15038-1000 15038-5000
TS15038-0250EA 80 USD
TS15038-0250 TS15038-1000 TS15038-5000
2-Methylbenzothiazole 99%
2-Methylbenzothiazole
Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: 12…14 °C
Density: 1.2 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00005794
CAS Number: 120-75-2
EINECS: 204-423-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6150 to 1.6180 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR