2-Picoline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylpyridine, α-Picoline

14933-0010 14933-0025 14933-0250
TS14933-0010EA 70 USD
TS14933-0010 TS14933-0025 TS14933-0250
2-Picoline 98%
2-Picoline
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 129.5
Melting Pt: -70
Density: 0.94 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00006332
CAS Number: 109-06-8
EINECS: 203-643-7
UN: 1992
ADR: 3 III
Merck Index: 12,07554

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4990 to 1.5010 (20°C, 589 nm)
Water ≤0.2 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR