2-Amino-6-chloropurine 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine

14929-0010 14929-0050
TS14929-0010EA 57 USD
TS14929-0010 TS14929-0050
2-Amino-6-chloropurine 99+%
2-Amino-6-chloropurine
Formula: C₅H₄ClN₅
MW: 169.57 g/mol
Melting Pt: 300
MDL Number: MFCD00075252
CAS Number: 10310-21-1
EINECS: 233-686-7

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
HPLC ≥99.0 %
Titration with HClO4 ≥99.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR