2-Aminobenzothiazole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II

14881-0050 14881-1000 14881-5000
TS14881-0050EA 24 USD
TS14881-0050 TS14881-1000 TS14881-5000
2-Aminobenzothiazole 97%
2-Aminobenzothiazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
Boiling Pt: 190…195 °C (0.05 mmHg)
Melting Pt: 129…134 °C
MDL Number: MFCD00005785
CAS Number: 136-95-8
EINECS: 205-268-4
Merck Index: 13,00423

Order Now

Specification Test Results

Appearance Beige to grey Powder or flakes
Infrared spectrum Conforms
Melting point 129 to 134 °C
Water ≤1 %
HPLC ≥97.0 %
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR