2,5-Dibromothiophene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14868-0250
14868-1000
14868-5000
TS14868-0250EA
45
USD
TS14868-0250
TS14868-1000
TS14868-5000
2,5-Dibromothiophene 95%
2,5-Dibromothiophene
Formula:
C₄H₂Br₂S Boiling Pt: 211 °C (760 mmHg) Melting Pt: –6 °C Density: 2.14 g/cm³ Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005420 CAS Number: 3141-27-3 EINECS: 221-547-3 |
Specification Test Results
Appearance | Clear light yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6270 to 1.6300 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...