Bis (8-hydroxyquinolinium) sulfate monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

14845-0025 14845-1000 14845-5000
TS14845-0025EA 905 USD
TS14845-0025 TS14845-1000 TS14845-5000
Bis (8-hydroxyquinolinium) sulfate monohydrate 98%
Bis(8-hydroxyquinolinium)sulphate monohydrate
Formula: C₁₈H₁₆N₂O₆S·1H₂O
Melting Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00149610
CAS Number: 207386-91-2
EINECS: 205-137-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder and/or lumps
Infrared spectrum Conforms
Water ≤4.5 %
Titration with CHA ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR