3-Hydroxy-2-nitropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol

14810-0250 14810-1000
TS14810-0250EA 79 USD
TS14810-0250 TS14810-1000
3-Hydroxy-2-nitropyridine 97%
3-Hydroxy-2-nitropyridine
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Melting Pt: 67…72 °C
MDL Number: MFCD00006258
CAS Number: 15128-82-2
EINECS: 239-191-2

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 72 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR