3-Amino-9-ethyl carbazole (AEC) 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9-Ethylcarbazol-3-ylamine, 9-Ethyl-9H-carbazol-3-amine, AEC, 50X, AEC, 3-Amino-9-ethylcarbazole

14787-0050 14787-0250 14787-1000
TS14787-0050EA 24 USD
TS14787-0050 TS14787-0250 TS14787-1000
3-Amino-9-ethyl carbazole (AEC) 90% tech.
3-Amino-9-ethyl carbazole (AEC)
Formula: C₁₄H₁₄N₂
MW: 210.28 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004964
CAS Number: 132-32-1
EINECS: 205-057-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder and/or chunks
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR