Nicotinic acid N-oxide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxiniacic Acid, 3-Carboxypyridine 1-oxide

14778-0050 14778-1000 14778-5000
TS14778-0050EA 18 USD
TS14778-0050 TS14778-1000 TS14778-5000
Nicotinic acid N-oxide 99%
Nicotinic acid N-oxide
Formula: C₆H₅NO₃
Melting Pt: 260…262 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006201
CAS Number: 2398-81-4

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH 98.5 to 101.5 %
Specific optical rotation -38.0° to -44.0° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR