Benzothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzo[d]thiazole

14639-0010 14639-0025 14639-0050 14639-2500
TS14639-0010EA 230 USD
TS14639-0010 TS14639-0025 TS14639-0050 TS14639-2500
Benzothiazole 97%
Benzothiazole
Formula: C₇H₅NS
Boiling Pt: 231 °C (760 mmHg)
Melting Pt: 2 °C
Density: 1.23 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00005775
CAS Number: 95-16-9
EINECS: 202-396-2
UN: 2810
ADR: 6.1,III
Merck Index: 13,01108

Order Now

Specification Test Results

Appearance Clear yellow-brown to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6410 to 1.6440 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR