3-Amino-1,2,4-triazine 97%

Supplier: Thermo Scientific Chemicals
Warning

14623-0100 14623-0500
TS14623-0100EA 76 USD
TS14623-0100 TS14623-0500
3-Amino-1,2,4-triazine 97%
3-Amino-1,2,4-triazine
Formula: C₃H₄N₄
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00006459
CAS Number: 1120-99-6
EINECS: 214-324-7

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 178 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR