4-Vinylpyridine 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger

14091-0010 14091-1000 14091-2500
TS14091-0010EA 283 USD
TS14091-0010 TS14091-1000 TS14091-2500
4-Vinylpyridine 95% stabilized
4-Vinylpyridine
Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 62…65 °C (15 mmHg)
Melting Pt: –20 °C
Density: 0.989 g/cm³
Flash Pt: 48 °C
Storage Temperature: Freezer
MDL Number: MFCD00006447
CAS Number: 100-43-6
EINECS: 202-852-0
UN: 3073
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to red-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5540 (20°C, 589 nm)
GC ≥94.0 %
Stabilizer 80 to 150 ppm Hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR