(±)-γ-Valerolactone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone

14079-1000 14079-5000
TS14079-0050EA 29.67 USD
TS14079-0050 TS14079-1000 TS14079-5000
(±)-γ-Valerolactone 98%
(±)-γ-Valerolactone
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 207…208 °C (760 mmHg)
Melting Pt: –31 °C
Density: 1.05 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005400
CAS Number: 108-29-2
EINECS: 203-569-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4340 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR