Trithiocyanuric acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14053-0050
14053-0250
TS14053-0050EA
46
USD
TS14053-0050
TS14053-0250
Trithiocyanuric acid 95%
Trithiocyanuric acid
Formula:
C₃H₃N₃S₃ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006052 CAS Number: 638-16-4 EINECS: 211-322-8 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...