1,3,5-Trithiane

Supplier: Thermo Scientific Chemicals
Danger

14052-0250 14052-1000
TS14052-0250EA 80 USD
TS14052-0250 TS14052-1000
1,3,5-Trithiane
1,3,5-Trithiane
Formula: C₃H₆S₃
Melting Pt: 214…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00006653
CAS Number: 291-21-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Carbon (C) 25.1 to 27.0
Sulfur (S) 67.1 to 72.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR