2,3,3-Trimethylindolenine 98%

Supplier: Thermo Scientific Chemicals
Warning

14016-0250 14016-1000 14016-2500
TS14016-0250EA 68 USD
TS14016-0250 TS14016-1000 TS14016-2500
2,3,3-Trimethylindolenine 98%
2,3,3-Trimethylindolenine
Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 228…229 °C (744 mmHg)
Density: 0.99 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005724
CAS Number: 1640-39-7
EINECS: 216-685-6

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR