2,4,6-Triaminopyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrimidine-2,4,6-triyltriamine

13925-0250 13925-1000
TS13925-0250EA 64 USD
TS13925-0250 TS13925-1000
2,4,6-Triaminopyrimidine 97%
2,4,6-Triaminopyrimidine
Formula: C₄H₇N₅
MW: 125.13 g/mol
Melting Pt: 246…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00006100
CAS Number: 1004-38-2
EINECS: 213-720-7

Order Now

Specification Test Results

Appearance Almost white to yellow to light brown Powder
Infrared spectrum Conforms
Melting point 246 to 252 °C
Titration with HClO4 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR