2-Thiophenemethanol 99+%

Supplier: Thermo Scientific Chemicals
Warning

13886-0250 13886-1000
TS13886-0250EA 65 USD
TS13886-0250 TS13886-1000
2-Thiophenemethanol 99+%
2-Thiophenemethanol
Formula: C₅H₆OS
Boiling Pt: 206…207 °C (761 mmHg)
Density: 0.9 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005454
CAS Number: 636-72-6
EINECS: 211-264-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5660 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR