Thiophene-2-carboxylic hydrazide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thiophenecarboxylic hydrazide, 2-Thiophenecarboxylic acid hydrazide, Thiophene-2-carbohydrazide

13885-0050 13885-0250
TS13885-0050EA 52 USD
TS13885-0050 TS13885-0250
Thiophene-2-carboxylic hydrazide 97%
Thiophene-2-carboxylic hydrazide
Formula: C₅H₆N₂OS
Melting Pt: 136…139 °C
MDL Number: MFCD00005435
CAS Number: 2361-27-5
EINECS: 219-107-0

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR