2-Thenoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiophene-2-carboxylic acid, 2-Thiophenecarboxylic acid

13884-0050 13884-0250 13884-1000 13884-5000
TS13884-0050EA 23 USD
TS13884-0050 TS13884-0250 TS13884-1000 TS13884-5000
2-Thenoic acid 99%
2-Thenoic acid
Formula: C₅H₄O₂S
Boiling Pt: 260 °C (760 mmHg)
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00005437
CAS Number: 527-72-0
EINECS: 208-423-4
Merck Index: 12,09491

Order Now

Specification Test Results

Appearance White to cream to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 130 °C
GC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR