2-Thiohydantoin 98%

Supplier: Thermo Scientific Chemicals
Warning

13876-0050 13876-0250 13876-1000
TS13876-0050EA 32 USD
TS13876-0050 TS13876-0250 TS13876-1000
2-Thiohydantoin 98%
2-Thiohydantoin
Formula: C₃H₄N₂OS
Melting Pt: 229…231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005277
CAS Number: 503-87-7
EINECS: 207-977-4

Order Now

Specification Test Results

Appearance Cream to yellow to beige to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR