(±)-Tetrahydrofurfuryl alcohol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA

13818-0010 13818-0025
TS13818-0010EA 103 USD
TS13818-0010 TS13818-0025
(±)-Tetrahydrofurfuryl alcohol 98%
(±)-Tetrahydrofurfuryl alcohol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –80 °C
Density: 1.0543 g/cm³
Flash Pt: 74 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
Water ≤0.3 %
GC ≥97.5 %
Color scale ≤50 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR