4-(1-Pyrrolidinyl)piperidine 99%

Supplier: Thermo Scientific Chemicals
Warning

13701-0010 13701-0050
TS13701-0010EA 45 USD
TS13701-0010 TS13701-0050
4-(1-Pyrrolidinyl)piperidine 99%
4-(1-Pyrrolidinyl)piperidine
Formula: C₉H₁₈N₂
MW: 154.2526 g/mol
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00038011
CAS Number: 5004-07-9

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥98.5 %
Specific optical rotation +240° to +260° (20 °C, 589 nm) c=0.5, methanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR