Phenyl glycidyl ether 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glycidyl phenyl ether

13535-0500 13535-2500
TS13535-0500EA 38 USD
TS13535-0500 TS13535-2500
Phenyl glycidyl ether 99%
Phenyl glycidyl ether
Formula: C₉H₁₀O₂
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: 3.5 °C
Density: 1.1 g/cm³
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00005133
CAS Number: 122-60-1
EINECS: 204-557-2
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5295 to 1.5315 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR