1-(2-Pyrimidyl)piperazine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(1-Piperazinyl)pyrimidine, 2-(Piperazin-1-yl)pyrimidine

13432-0010 13432-0050
TS13432-0010EA 23 USD
TS13432-0010 TS13432-0050 TS13432-0250
1-(2-Pyrimidyl)piperazine 98+%
1-(2-Pyrimidyl)piperazine
Formula: C₈H₁₂N₄
Boiling Pt: 96…97 °C (.2 mmHg)
Melting Pt: 29…36 °C
Density: 1.07 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040742
CAS Number: 20980-22-7
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5850 to 1.5880 (20°C, 589 nm)
GC ≥98.0 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR