4-Methylindole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13389-0010
13389-0050
TS13389-0010EA
387
USD
TS13389-0010
TS13389-0050
4-Methylindole 99%
4-Methylindole
Formula:
C₉H₉N Boiling Pt: 267 °C (760 mmHg) Melting Pt: 5 °C Density: 1.06 g/cm³ Flash Pt: >112 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005668 CAS Number: 16096-32-5 EINECS: 240-262-5 UN: 3334 ADR: 9,III |
Specification Test Results
Appearance | Clear yellow to brown Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.6060 to 1.6080 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...