3-Aminoisoxazole 95%

Supplier: Thermo Scientific Chemicals
Warning

13353-0050 13353-0250
TS13353-0050EA 119 USD
TS13353-0050 TS13353-0250
3-Aminoisoxazole 95%
3-Aminoisoxazole
Formula: C₃H₄N₂O
MW: 84.08 g/mol
Boiling Pt: 226…228 °C (760 mmHg)
Density: 1.38 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00038814
CAS Number: 1750-42-1

Order Now

Specification Test Results

Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5090 to 1.5120 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR