trans-Stilbene oxide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-2,3-Diphenylethylene Oxide

13278-0010 13278-0050 13278-0250
TS13278-0010EA 32 USD
TS13278-0010 TS13278-0050 TS13278-0250
trans-Stilbene oxide 99%
trans-Stilbene oxide
Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00064311
CAS Number: 1439-07-2

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR