Sesamol-(3,4-(methylenedioxy)phenol) 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Sesamol, 3,4-Methylenedioxyphenol, Benzo[d][1,3]dioxol-5-ol
13269-0050
13269-0250
13269-1000
TS13269-0050EA
32
USD
TS13269-0050
TS13269-0250
TS13269-1000
Sesamol-(3,4-(methylenedioxy)phenol) 98%
Sesamol-(3,4-(methylenedioxy)phenol)
Formula:
C₇H₆O₃ MW: 138.12 g/mol Melting Pt: 63…65 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005827 CAS Number: 533-31-3 EINECS: 208-561-5 |
Specification Test Results
Appearance | Off-white to beige Crystalline powder (May darken on storage) |
Infrared spectrum | Conforms |
Melting point | 63 to 65 °C |
Water | ≤0.2 % (K.F.) |
HPLC | ≥97.5 % |
Specific optical rotation | +80.0° to +90.0° (20 °C, 589 nm) (c=1, water, after 5 hours) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...