Quinoline 96%

Supplier: Thermo Scientific Chemicals
Danger

13219-0025 13219-0100 13219-5000
TS13219-0010EA 141 USD
TS13219-0010 TS13219-0025 TS13219-0100 TS13219-5000
Quinoline 96%
Quinoline
Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 237 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1.095 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006736
CAS Number: 91-22-5
EINECS: 202-051-6
UN: 2656
ADR: 6.1,III
Merck Index: 13,08160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e452fdc6c4f70d31bc2cfb5dade041787f8be427&vid=ec752804bedb2e9f8adfbea9ad287c2452f87a17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear light yellow to dark brown Liquid
Infrared spectrum Conforms
Refractive index 1.6230 to 1.6270 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR