Quinaldine 95+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylquinoline

13217-0010 13217-2500
TS13217-0010EA 179 USD
TS13217-0010 TS13217-2500
Quinaldine 95+%
Quinaldine
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 248 °C (760 mmHg)
Melting Pt: –2 °C
Density: 1.058 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00006756
CAS Number: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228

Order Now

Specification Test Results

Appearance Clear light yellow to brown-yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6080 to 1.6130 (20°C, 589 nm)
GC ≥95.0 %
Hydroxyl value 25 to 32 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR