Pyridine-3,5-dicarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dinicotinic acid, 3,5-Pyridinedicarboxylic acid
13190-0100
13190-0500
TS13190-0100EA
101
USD
TS13190-0100
TS13190-0500
Pyridine-3,5-dicarboxylic acid 98%
Pyridine-3,5-dicarboxylic acid
Formula:
C₇H₅NO₄ MW: 167.12 g/mol Boiling Pt: 357 °C (1013 hPa) Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006393 CAS Number: 499-81-0 EINECS: 207-893-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...