Pyridine-3,4-dicarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Pyridinedicarboxylic acid, Cinchomeronic acid, Pyridine-3,4-dicarboxylicacid

13189-0050 13189-0250
TS13189-0050EA 24 USD
TS13189-0050 TS13189-0250
Pyridine-3,4-dicarboxylic acid 97%
Pyridine-3,4-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 529 °C (1013 hPa)
Melting Pt: 262 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006392
CAS Number: 490-11-9
EINECS: 207-705-4
Merck Index: 13,02304

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR