Pyridine-2,6-dicarboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA

13188-0250 13188-1000 13188-5000
TS13188-0250EA 39 USD
TS13188-0250 TS13188-1000 TS13188-5000
Pyridine-2,6-dicarboxylic acid 99%
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
Melting Pt: 327
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.3 % (K.F.)
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR