syn-Pyridine-2-carboxaldoxime 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: syn-2-Pyridinealdoxime

13179-0250 13179-1000
TS13179-0250EA 91 USD
TS13179-0250 TS13179-1000
syn-Pyridine-2-carboxaldoxime 99+%
syn-Pyridine-2-carboxaldoxime
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00167166
CAS Number: 1193-96-0

Order Now

Specification Test Results

Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 113 °C
Water ≤0.2 % (K.F.)
Titration with HClO4 99.0 to 101.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR