Pyridazine 98+%

Supplier: Thermo Scientific Chemicals

13176-0050 13176-0500
TS13176-0050EA 70 USD
TS13176-0050 TS13176-0500
Pyridazine 98+%
Pyridazine
Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 208 °C (760 mmHg)
Melting Pt: –8 °C
Density: 1.103 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00006463
CAS Number: 289-80-5
EINECS: 206-025-5
Merck Index: 12,08152

Order Now

Specification Test Results

Appearance Clear yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5225 to 1.5245 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR