Pyrazine 99+%

Supplier: Thermo Scientific Chemicals
Warning

13170-0100 13170-0250 13170-1000
TS13170-0100EA 46 USD
TS13170-0100 TS13170-0250 TS13170-1000
Pyrazine 99+%
Pyrazine
Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 115…116 °C (760 mmHg)
Melting Pt: 52…56 °C
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00006122
CAS Number: 290-37-9
EINECS: 206-027-6
UN: 1325
ADR: 4.1,III
Merck Index: 13,08047

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 52 to 56 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR