Piperonylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

13137-0050 13137-0250 13137-1000
TS13137-0050EA 42 USD
TS13137-0050 TS13137-0250 TS13137-1000
Piperonylic acid 99%
Piperonylic acid
Formula: C₈H₆O₄
Melting Pt: 229…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00005830
CAS Number: 94-53-1
EINECS: 202-342-8
Merck Index: 12,07630

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 229 to 235 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR