Picolinic acid N-oxide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13118-0050
13118-0250
TS13118-0050EA
36
USD
TS13118-0050
TS13118-0250
Picolinic acid N-oxide 97%
Picolinic acid N-oxide
Formula:
C₆H₅NO₃ MW: 139.1088 g/mol Melting Pt: >150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006198 CAS Number: 824-40-8 EINECS: 212-528-0 |
Specification Test Results
Appearance | White to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | ≥145 °C |
Water | ≤0.5 % (K.F.) |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...