3-Picoline 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: β-Picoline, 3-Methylpyridine

13113-0010 13113-0025 13113-2500 13113-5000
TS13113-0010EA 97 USD
TS13113-0010 TS13113-0025 TS13113-2500 TS13113-5000
3-Picoline 99%
3-Picoline
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 144
Melting Pt: -19.4
Density: 0.95 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00006402
CAS Number: 108-99-6
EINECS: 203-636-9
UN: 2313
ADR: 3 III
Merck Index: 13,07485

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5050 to 1.5070 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR