4-Phenylpyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

13091-0050 13091-0250 13091-1000
TS13091-0050EA 79 USD
TS13091-0050 TS13091-0250 TS13091-1000
4-Phenylpyridine 99%
4-Phenylpyridine
Formula: C₁₁H₉N
Boiling Pt: 274…275 °C (760 mmHg)
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00006420
CAS Number: 939-23-1
EINECS: 213-357-4

Order Now

Specification Test Results

Appearance White to light yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 71 to 75 °C
GC ≥98.5 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR