2-Phenylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

13090-0050 13090-0250
TS13090-0050EA 44 USD
TS13090-0050 TS13090-0250
2-Phenylpyridine 97%
2-Phenylpyridine
Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 268…270 °C (760 mmHg)
Density: 1.086 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6220 to 1.6260 (20°C, 589 nm)
GC ≥96.0 %
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR