N-Phenylmaleimide 97%

Supplier: Thermo Scientific Chemicals
Danger

13071-0250 13071-1000
TS13071-0250EA 66 USD
TS13071-0250 TS13071-1000
N-Phenylmaleimide 97%
N-Phenylmaleimide
Formula: C₁₀H₇NO₂
Boiling Pt: 162…163 °C (12 mmHg)
Melting Pt: 86…90 °C
MDL Number: MFCD00005502
CAS Number: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,07452

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 86 to 91 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR