Phenazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13015-0050
13015-0100
TS13015-0050EA
47
USD
TS13015-0050
TS13015-0100
Phenazine 98%
Phenazine
Formula:
C₁₂H₈N₂ MW: 180.21 g/mol Boiling Pt: >360 °C (760 mmHg) Melting Pt: 174…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005023 CAS Number: 92-82-0 EINECS: 202-193-9 Merck Index: 12,07359 |
Specification Test Results
Appearance | Yellow to brown Crystalline powder or crystals or needles |
Infrared spectrum | Conforms |
Melting point | 173 to 177 °C |
HPLC | ≥97.5 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.5 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...